3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione ≥98.0%
Supplier: TCI America
Synonyms:
3,6-Diphenylpyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione, Pigment Red 255
D3743-5G
D3743-25G
TCD3743-5GEA
169.06
CAD
TCD3743-5G
TCD3743-25G
3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione ≥98.0%
3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione
CAS Number: 54660-00-3
Molecular Formula: C18H12N2O2
Molecular Weight: 288.31
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 360
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: Pigment Red 255
Formula:
C₁₈H₁₂N₂O₂ Melting Pt: 360 °C |
MDL Number:
MFCD11870113 CAS Number: 54660-00-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...