N-[2-(Dimethylamino)ethyl]-N-methylformamide ≥97.0% (by GC, titration analysis)
Supplier: TCI America
D3724-5G
D3724-25G
TCD3724-5GEA
109.02
CAD
TCD3724-5G
TCD3724-25G
N-[2-(Dimethylamino)ethyl]-N-methylformamide ≥97.0% (by GC, titration analysis)
N-[2-(Dimethylamino)ethyl]-N-methylformamide
CAS Number: 105669-53-2
MDL Number: MFCD04039895
Molecular Formula: C6H14N2O
Molecular Weight: 130.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 70
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: N-Formyl-N,N',N'-trimethylethylenediamine
Formula:
C₆H₁₄N₂O Boiling Pt: 96 °C Density: 0.95 (20 °C) Flash Pt: 70 °C |
MDL Number:
MFCD04039895 CAS Number: 105669-53-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...