2-Amino-4'-chlorobenzophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (2-Aminophenyl)(4-chlorophenyl)methanone

A1244-25G A1244-5G
TCA1244-025GEA 252.96 CAD
TCA1244-025G TCA1244-5G
2-Amino-4'-chlorobenzophenone ≥98.0% (by GC, titration analysis)
2-Amino-4'-chlorobenzophenone

CAS Number: 2894-51-1
MDL Number: MFCD00025193
Molecular Formula: C13H10ClNO
Molecular Weight: 231.68
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 103

Formula: C₁₃H₁₀ClNO
MW: 231.6800 g/mol
Melting Pt: 103 °C
MDL Number: MFCD00025193
CAS Number: 2894-51-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd405e20af9cef2e44ddf439e268895eb6888861&vid=dd405e20af9cef2e44ddf439e268895eb6888861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR