9-Decyn-1-ol ≥94.0%

Supplier: TCI America

Synonyms: Dec-9-yn-1-ol, 10-Hydroxy-1-decyne

D3710-5G D3710-25G
TCD3710-5GEA 213.3 CAD
TCD3710-5G TCD3710-25G
9-Decyn-1-ol ≥94.0%
9-Decyn-1-ol

CAS Number: 17643-36-6
MDL Number: MFCD00049199
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Specific Gravity (20/20): 0.87


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 10-Hydroxy-1-decyne

Formula: C₁₀H₁₈O
Boiling Pt: 92 °C
Density: 0.87 (20 °C)
MDL Number: MFCD00049199
CAS Number: 17643-36-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd29dc31e508525be478d3933990f929a917d68f&vid=fd29dc31e508525be478d3933990f929a917d68f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR