Diethyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate ≥97.0%

Supplier: TCI America

D3708-5G D3708-1G
TCD3708-5GEA 176.8 CAD
TCD3708-5G TCD3708-1G
Diethyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate ≥97.0%
Diethyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate

CAS Number: 124931-12-0
MDL Number: MFCD00134233
Molecular Formula: C8H18NO5P
Molecular Weight: 239.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 110
Specific Gravity (20/20): 1.16


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (N-Methoxy-N-methylcarbamoylmethyl)phosphonic Acid Diethyl Ester

Formula: C₈H₁₈NO₅P
Boiling Pt: 124 °C
Density: 1.16 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00134233
CAS Number: 124931-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d37022261b47865107db721e6d59efbe2ba14ce4&vid=d37022261b47865107db721e6d59efbe2ba14ce4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR