Dimethyl-cis-cyclohexane-1,2-dicarboxylate ≥96.0%

Supplier: TCI America

D3702-5G D3702-1G
TCD3702-5GEA 495.8 CAD
TCD3702-5G TCD3702-1G
Dimethyl-cis-cyclohexane-1,2-dicarboxylate ≥96.0%
Dimethyl cis-cyclohexane-1,2-dicarboxylate

CAS Number: 1687-29-2
MDL Number: MFCD00043768
Molecular Formula: C10H16O4
Molecular Weight: 200.23
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 111
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: cis-1,2-Cyclohexanedicarboxylic Acid Dimethyl Ester

Formula: C₁₀H₁₆O₄
Boiling Pt: 111 °C
Density: 1.12 (20 °C)
MDL Number: MFCD00043768
CAS Number: 1687-29-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb54dde4766bd133885bb8954d9bcbe34a8364e5&vid=bb54dde4766bd133885bb8954d9bcbe34a8364e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR