4,6-Dimethyl-2-methylsulfonylpyrimidine ≥98.0%
Supplier: TCI America
D3699-25G
D3699-5G
TCD3699-25GEA
426.6
CAD
TCD3699-25G
TCD3699-5G
4,6-Dimethyl-2-methylsulfonylpyrimidine ≥98.0%
4,6-Dimethyl-2-methylsulfonylpyrimidine
CAS Number: 35144-22-0
MDL Number: MFCD03788286
Molecular Formula: C7H10N2O2S
Molecular Weight: 186.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82
Formula:
C₇H₁₀N₂O₂S MW: 186.23 g/mol Melting Pt: 82 °C |
MDL Number:
MFCD03788286 CAS Number: 35144-22-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...