3,5-Dimethyl-1-adamantanol ≥97.0%
Supplier: TCI America
D3697-5G
D3697-1G
TCD3697-5GEA
455.04
CAD
TCD3697-5G
TCD3697-1G
3,5-Dimethyl-1-adamantanol ≥97.0%
3,5-Dimethyl-1-adamantanol
CAS Number: 707-37-9
MDL Number: MFCD00074775
Molecular Formula: C12H20O
Molecular Weight: 180.29
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 95
Other Product Names / Synonyms: 1-Hydroxy-3,5-dimethyladamantane
Formula:
C₁₂H₂₀O Melting Pt: 95 °C |
MDL Number:
MFCD00074775 CAS Number: 707-37-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...