Dimethyl-trans-4-cyclohexene-1,2-dicarboxylate ≥98.0%

Supplier: TCI America

D3692-1G D3692-5G
TCD3692-1GEA 243 CAD
TCD3692-1G TCD3692-5G
Dimethyl-trans-4-cyclohexene-1,2-dicarboxylate ≥98.0%
Dimethyl-trans-4-cyclohexene-1,2-dicarboxylate

CAS Number: 17673-68-6
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 119
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: trans-4-Cyclohexene-1,2-dicarboxylic Acid Dimethyl Ester

Formula: C₁₀H₁₄O₄
Boiling Pt: 119 °C
Density: 1.13 (20 °C)
CAS Number: 17673-68-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b5a4b7edba6fad9b82cb32ba29c5dc924a560e7&vid=8b5a4b7edba6fad9b82cb32ba29c5dc924a560e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR