Diethyl (4-iodobenzyl)phosphonate ≥98.0%

Supplier: TCI America

Synonyms: Diethyl 4-iodobenzylphosphonate, (4-Iodobenzyl)phosphonic acid diethyl ester

D3689-25G D3689-5G
TCD3689-25GEA 710.68 CAD
TCD3689-25G TCD3689-5G
Diethyl (4-iodobenzyl)phosphonate ≥98.0%
Diethyl (4-iodobenzyl)phosphonate

CAS Number: 173443-43-1
MDL Number: MFCD09033365
Molecular Formula: C11H16IO3P
Molecular Weight: 354.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.53


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (4-Iodobenzyl)phosphonic Acid Diethyl Ester

Formula: C₁₁H₁₆IO₃P
MW: 354.121 g/mol
Density: 1.53 (20 °C)
MDL Number: MFCD09033365
CAS Number: 173443-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd865cb8feb4786a324ab31a87b19d1fb4f3d824&vid=bd865cb8feb4786a324ab31a87b19d1fb4f3d824&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR