Diethyl (4-bromobenzyl)phosphonate ≥98.0%

Supplier: TCI America

D3688-25G D3688-5G
TCD3688-25GEA 257.54 CAD
TCD3688-25G TCD3688-5G
Diethyl (4-bromobenzyl)phosphonate ≥98.0%
Diethyl (4-bromobenzyl)phosphonate

CAS Number: 38186-51-5
MDL Number: MFCD05663693
Molecular Formula: C11H16BrO3P
Molecular Weight: 307.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Specific Gravity (20/20): 1.37


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (4-Bromobenzyl)phosphonic Acid Diethyl Ester

Formula: C₁₁H₁₆BrO₃P
Boiling Pt: 183 °C
Density: 1.37 (20 °C)
MDL Number: MFCD05663693
CAS Number: 38186-51-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad2b1235b740340f05ed86c21dbb3282cbc8392e&vid=ad2b1235b740340f05ed86c21dbb3282cbc8392e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR