Diphenylammonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Diphenylammonium Trifluoromethanesulphonate, DPAT, Diphenylammonium Trifluoromethanesulfonate

D3683-25G D3683-1G D3683-5G
TCD3683-25GEA 816.86 CAD
TCD3683-25G TCD3683-1G TCD3683-5G
Diphenylammonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)
Diphenylammonium trifluoromethanesulfonate

CAS Number: 164411-06-7
Molecular Formula: C13H12F3NO3S
Molecular Weight: 319.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification
Other Product Names / Synonyms: DPAT

Formula: C₁₃H₁₂F₃NO₃S
MW: 319.30 g/mol
MDL Number: MFCD11042671
CAS Number: 164411-06-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=894cee608cc8570c54d19bfd7bcdbfadb9d3666e&vid=894cee608cc8570c54d19bfd7bcdbfadb9d3666e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR