Dimethyl3,4-dihydroxy-2,5-thiophenedicarboxylate ≥98.0%

Supplier: TCI America

D3682-1G D3682-5G
TCD3682-1GEA 185.18 CAD
TCD3682-1G TCD3682-5G
Dimethyl3,4-dihydroxy-2,5-thiophenedicarboxylate ≥98.0%
Dimethyl3,4-dihydroxy-2,5-thiophenedicarboxylate

CAS Number: 58416-04-9
MDL Number: MFCD01860087
Molecular Formula: C8H8O6S
Molecular Weight: 232.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 182


Other Product Names / Synonyms: 3,4-Dihydroxy-2,5-thiophenedicarboxylic Acid Dimethyl Ester

Formula: C₈H₈O₆S
Melting Pt: 182 °C
MDL Number: MFCD01860087
CAS Number: 58416-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42a63e802f2727c7642a8bbac395e3e88e16c542&vid=42a63e802f2727c7642a8bbac395e3e88e16c542&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR