2,6-Dimethylbenzaldehyde ≥97.0%

Supplier: TCI America

D3681-25G D3681-5G
TCD3681-25GEA 475.58 CAD
TCD3681-25G TCD3681-5G
2,6-Dimethylbenzaldehyde ≥97.0%
2,6-Dimethylbenzaldehyde

CAS Number: 1123-56-4
MDL Number: MFCD00128003
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 230
Flash Point (°C): 96
Freezing point (°C): 27


Other Product Names / Synonyms: m-Xylene-2-carboxaldehyde

Formula: C₉H₁₀O
Boiling Pt: 230 °C
Flash Pt: 96 °C
MDL Number: MFCD00128003
CAS Number: 1123-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b87e0b6ceed0a727ad630a118f0c60f6b55a4c38&vid=b87e0b6ceed0a727ad630a118f0c60f6b55a4c38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR