2,5-Dichloro-3-methylthiophene ≥96.0%

Supplier: TCI America

D3680-1G D3680-5G
TCD3680-1GEA 52.93 CAD
TCD3680-1G TCD3680-5G
2,5-Dichloro-3-methylthiophene ≥96.0%
2,5-Dichloro-3-methylthiophene

CAS Number: 17249-90-0
MDL Number: MFCD00022490
Molecular Formula: C5H4Cl2S
Molecular Weight: 167.05
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Flash Point (°C): 77
Specific Gravity (20/20): 1.37
Storage Temperature: 0-10°C

Formula: C₅H₄Cl₂S
MW: 167.06 g/mol
Boiling Pt: 185 °C
Density: 1.37 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00022490
CAS Number: 17249-90-0
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff6fd7fd43f696c21cbab49aac0b1cd053327d69&vid=ff6fd7fd43f696c21cbab49aac0b1cd053327d69&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR