3,6-Dibromothieno[3,2-b]thiophene ≥98.0%
Supplier: TCI America
D3679-1G
D3679-200MG
TCD3679-1GEA
300.83
CAD
TCD3679-1G
TCD3679-200MG
3,6-Dibromothieno[3,2-b]thiophene ≥98.0%
3,6-Dibromothieno[3,2-b]thiophene
CAS Number: 392662-65-6
Molecular Formula: C6H2Br2S2
Molecular Weight: 298.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 127
Storage Temperature: 0-10°C
Formula:
C₆H₂Br₂S₂ Melting Pt: 127 °C |
MDL Number:
MFCD10000958 CAS Number: 392662-65-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...