2,5-Dibromothieno[3,2-b]thiophene ≥98.0%
Supplier: TCI America
D3678-200MG
D3678-5G
D3678-1G
TCD3678-200MGEA
75.84
CAD
TCD3678-200MG
TCD3678-5G
TCD3678-1G
2,5-Dibromothieno[3,2-b]thiophene ≥98.0%
2,5-Dibromothieno[3,2-b]thiophene
CAS Number: 25121-87-3
MDL Number: MFCD03931293
Molecular Formula: C6H2Br2S2
Molecular Weight: 298.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 130
Storage Temperature: <0°C
Formula:
C₆H₂Br₂S₂ Melting Pt: 130 °C |
MDL Number:
MFCD03931293 CAS Number: 25121-87-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...