2'-Deoxycytidine-5'-monophosphate hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D3673-100MG D3673-1G
TCD3673-100MGEA 69.36 CAD
TCD3673-100MG TCD3673-1G
2'-Deoxycytidine-5'-monophosphate hydrate ≥98.0% (by HPLC, titration analysis)
2'-Deoxycytidine-5'-monophosphate hydrate

CAS Number: 1032-65-1
Molecular Formula: C9H14N3O7P
Molecular Weight: 307.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 161
Specific rotation [a]20/D: 37 deg (C=1, H2O)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Nucleotides and Their Analogs
Other Product Names / Synonyms: dCMP, Deoxycytidylic Acid

Formula: C₉H₁₄N₃O₇P
Melting Pt: 161 °C
CAS Number: 1032-65-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e4aafeff5a24880615b7ff63336a50f016da292&vid=c0b9a2f010ac9bebe7af6adb6f98cf3e926044bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR