1,2-Dimethylbenzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3665-1G D3665-5G
TCD3665-1GEA 120.08 CAD
TCD3665-1G TCD3665-5G
1,2-Dimethylbenzimidazole ≥98.0% (by GC, titration analysis)
1,2-Dimethylbenzimidazole

CAS Number: 2876-08-6
MDL Number: MFCD00022840
Molecular Formula: C9H10N2
Molecular Weight: 146.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 164
Melting point (°C): 111

Formula: C₉H₁₀N₂
Boiling Pt: 164 °C
Melting Pt: 111 °C
MDL Number: MFCD00022840
CAS Number: 2876-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab2363a0cacd68b188dd12af1b79622e6d3444e2&vid=ab2363a0cacd68b188dd12af1b79622e6d3444e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR