(2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one ≥96.0%

Supplier: TCI America

Synonyms: (2S,3S)-3-Hydroxy-2-(4-methoxyphenyl)-2,3-dihydrobenzo[b][1,4]thiazepin-4(5H)-one

D3661-25G D3661-5G
TCD3661-25GEA 170.32 CAD
TCD3661-25G TCD3661-5G
(2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one ≥96.0%
(2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one

CAS Number: 42399-49-5
MDL Number: MFCD01321316
Molecular Formula: C16H15NO3S
Molecular Weight: 301.36
Purity/Analysis Method: >96.0% (HPLC,N)
Form: Crystal
Melting point (°C): 204
Specific rotation [a]20/D: 114 deg (C=0.5, MeOH)

Formula: C₁₆H₁₅NO₃S
MW: 301.36 g/mol
Melting Pt: 204 °C
MDL Number: MFCD01321316
CAS Number: 42399-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a89ccab387ebccd524046cc802c61751cec0e53&vid=6a89ccab387ebccd524046cc802c61751cec0e53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR