Dithiothreitol (DTT, Cleland's reagent) ≥98.0%

Supplier: TCI America

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

D3647-1G D3647-5G
TCD3647-1GEA 85.32 CAD
TCD3647-1G TCD3647-5G
Dithiothreitol (DTT, Cleland's reagent) ≥98.0%
Dithiothreitol (DTT, Cleland's reagent)

CAS Number: 3483-12-3
MDL Number: MFCD00004877
Molecular Formula: C4H10O2S2
Molecular Weight: 154.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 43
Flash Point (°C): 113
Storage Temperature: 0-10°C


Product Categories: Bioscience, Disulfide Reducing Agents, Analytical Science, Reagents for Gel & Buffer Preparation, etc. (Reagents for Electrophoresis)
Other Product Names / Synonyms: Cleland's Reagent, DL-2,3-Dihydroxy-1,4-butanedithiol, DL-1,4-Dimercapto-2,3-butanediol, DL-DTT

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Boiling Pt: 130 °C
Melting Pt: 43 °C
Flash Pt: 113 °C
MDL Number: MFCD00004877
CAS Number: 3483-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3972330e17cd18d000b14e4e871b4189c0a59bb&vid=012512492f94776a2f94c72c997dbde60b137106&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR