3,6-Di(2-pyridyl)-1,2,4,5-tetrazine ≥98.0%

Supplier: TCI America

Synonyms: 3,6-Di(pyridin-2-yl)-1,2,4,5-tetrazine, 3,6-Bis(2-pyridyl)-1,2,4,5-tetrazine

D3640-5G D3640-1G
TCD3640-5GEA 512.39 CAD
TCD3640-5G TCD3640-1G
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine ≥98.0%
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine

CAS Number: 1671-87-0
MDL Number: MFCD00121717
Molecular Formula: C12H8N6
Molecular Weight: 236.24
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 225


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: 3,6-Bis(2-pyridyl)-1,2,4,5-tetrazine

Formula: C₁₂H₈N₆
MW: 236.24 g/mol
Melting Pt: 225 °C
MDL Number: MFCD00121717
CAS Number: 1671-87-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ea789564d50591ddea65865bfab444dfb22492c&vid=0ea789564d50591ddea65865bfab444dfb22492c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR