Diethyl-trans-1,4-cyclohexanedicarboxylate ≥98.0%
Supplier: TCI America
Synonyms:
trans-1,4-Cyclohexanedicarboxylic Acid Diethyl Ester
D3639-25G
D3639-5G
TCD3639-25GEA
676.08
CAD
TCD3639-25G
TCD3639-5G
Diethyl-trans-1,4-cyclohexanedicarboxylate ≥98.0%
Diethyl-trans-1,4-cyclohexanedicarboxylate
CAS Number: 19145-96-1
Molecular Formula: C12H20O4
Molecular Weight: 228.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 45
Other Product Names / Synonyms: trans-1,4-Cyclohexanedicarboxylic Acid Diethyl Ester
Formula:
C₁₂H₂₀O₄ MW: 228.29 g/mol Melting Pt: 45 °C |
CAS Number:
19145-96-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...