(±)-3-Aminoquinuclidine dihydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 3-Aminoquinuclidine dihydrochloride, 3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

A1236-5G A1236-25G
TCA1236-005GEA 145.36 CAD
TCA1236-005G TCA1236-025G
(±)-3-Aminoquinuclidine dihydrochloride ≥98.0%
(±)-3-Aminoquinuclidine dihydrochloride

CAS Number: 6530-09-2
MDL Number: MFCD00137395
Molecular Formula: C7H14N2
Molecular Weight: 199.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 3-Amino-1-azabicyclo[2.2.2]octane Dihydrochloride

Formula: C₇H₁₆Cl₂N₂
MW: 199.1213 g/mol
MDL Number: MFCD00137395
CAS Number: 6530-09-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0574e72f5289c12733951b8368038acfe2914382&vid=0574e72f5289c12733951b8368038acfe2914382&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR