2,6-Dimethylpyridin-4-ol ≥98.0%
Supplier: TCI America
Synonyms:
2,6-Dimethyl-4-pyridinol, 2,6-Dimethyl-4-hydroxypyridine
D3638-5G
D3638-1G
TCD3638-5GEA
314.42
CAD
TCD3638-5G
TCD3638-1G
2,6-Dimethylpyridin-4-ol ≥98.0%
2,6-Dimethylpyridin-4-ol
CAS Number: 13603-44-6
MDL Number: MFCD00956021
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 351
Melting point (°C): 230
Other Product Names / Synonyms: 2,6-Dimethyl-4-pyridinol
Formula:
C₇H₉NO MW: 123.1525 g/mol Boiling Pt: 351 °C Melting Pt: 230 °C |
MDL Number:
MFCD00956021 CAS Number: 13603-44-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...