Diethyl-2-ethyl-2-methylmalonate ≥97.0%

Supplier: TCI America

Synonyms: 2-Ethyl-2-methylmalonic Acid Diethyl Ester

D3630-25G D3630-5G
TCD3630-25GEA 348.71 CAD
TCD3630-25G TCD3630-5G
Diethyl-2-ethyl-2-methylmalonate ≥97.0%
Diethyl-2-ethyl-2-methylmalonate

CAS Number: 2049-70-9
MDL Number: MFCD02258541
Molecular Formula: C10H18O4
Molecular Weight: 202.25
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 79
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: 2-Ethyl-2-methylmalonic Acid Diethyl Ester

Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 79 °C
Density: 0.99 (20 °C)
MDL Number: MFCD02258541
CAS Number: 2049-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ff22f51fa6a0c63a7e05303e007a3e1c1a026ae&vid=5ff22f51fa6a0c63a7e05303e007a3e1c1a026ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR