3,5-Dimethyl-1-adamantanamine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Dimethyl-5-aminoadamantane hydrochloride, Memantine Hydrochloride, 1-Amino-3,5-dimethyladamantane Hydrochloride

D3608-5G D3608-25G
TCD3608-5GEA 55.3 CAD
TCD3608-5G TCD3608-25G
3,5-Dimethyl-1-adamantanamine hydrochloride ≥98.0%
3,5-Dimethyl-1-adamantanamine hydrochloride

CAS Number: 41100-52-1
MDL Number: MFCD00214336
Molecular Formula: C12H21N
Molecular Weight: 215.77
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 292


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 1-Amino-3,5-dimethyladamantane Hydrochloride, Memantine Hydrochloride

Formula: C₁₂H₂₁N
MW: 215.77 g/mol
Melting Pt: 292 °C
MDL Number: MFCD00214336
CAS Number: 41100-52-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56d6e658a0d5f08c32a98d1b5a09d6e207593797&vid=56d6e658a0d5f08c32a98d1b5a09d6e207593797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR