3,3'-Dibromo-4,4'-dimethoxybiphenyl ≥95.0%
Supplier: TCI America
D3603-1G
D3603-5G
TCD3603-1GEA
216.46
CAD
TCD3603-1G
TCD3603-5G
3,3'-Dibromo-4,4'-dimethoxybiphenyl ≥95.0%
3,3'-Dibromo-4,4'-dimethoxybiphenyl
CAS Number: 108989-36-2
Molecular Formula: C14H12Br2O2
Molecular Weight: 372.06
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 166
Formula:
C₁₄H₁₂Br₂O₂ Melting Pt: 166 °C |
MDL Number:
MFCD10000957 CAS Number: 108989-36-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...