3,3'-Dibromo-4,4'-biphenol ≥98.0%

Supplier: TCI America

Synonyms: 3,3'-Dibromo-4,4'-dihydroxybiphenyl, 3,3'-Dibromobiphenyl-4,4'-diol, 3,3'-Dibromo-[1,1'-biphenyl]-4,4'-diol

D3602-1G D3602-5G
TCD3602-1GEA 380.15 CAD
TCD3602-1G TCD3602-5G
3,3'-Dibromo-4,4'-biphenol ≥98.0%
3,3'-Dibromo-4,4'-biphenol

CAS Number: 189039-64-3
Molecular Formula: C12H8Br2O2
Molecular Weight: 344.00
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 122


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3,3'-Dibromobiphenyl-4,4'-diol, 3,3'-Dibromo-4,4'-dihydroxybiphenyl

Formula: C₁₂H₈Br₂O₂
MW: 344.00 g/mol
Melting Pt: 122 °C
MDL Number: MFCD10566892
CAS Number: 189039-64-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=504b489e2c1d250b7a448c5a47fd62f405b829e0&vid=504b489e2c1d250b7a448c5a47fd62f405b829e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR