N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥98.0%
Supplier: TCI America
Synonyms:
4-(Diethylamino)phenylboronic Acid Pinacol Ester, 2-[4-(Diethylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CAS Number: 920304-57-0
Molecular Formula: C16H26BNO2
Molecular Weight: 275.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-[4-(Diethylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(Diethylamino)phenylboronic Acid Pinacol Ester
Formula:
C₁₆H₂₆BNO₂ MW: 275.20 g/mol |
MDL Number:
MFCD10000956 CAS Number: 920304-57-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...