2,5-Dibenzylidenecyclopentanone ≥98.0% (by HPLC)

Supplier: TCI America

D3597-25G D3597-5G
TCD3597-25GEA 737.23 CAD
TCD3597-25G TCD3597-5G
2,5-Dibenzylidenecyclopentanone ≥98.0% (by HPLC)
2,5-Dibenzylidenecyclopentanone

CAS Number: 895-80-7
MDL Number: MFCD00019321
Molecular Formula: C19H16O
Molecular Weight: 260.34
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 192

Formula: C₁₉H₁₆O
Melting Pt: 192 °C
MDL Number: MFCD00019321
CAS Number: 895-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cad784c911e62266ba6efe40adec6bcd85fcdbd7&vid=cad784c911e62266ba6efe40adec6bcd85fcdbd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR