Dimethyl tetrafluorosuccinate ≥95.0%

Supplier: TCI America

Synonyms: Perfluorosuccinic Acid Dimethyl Ester, Tetrafluorobutanedioic Acid Dimethyl Ester, Dimethyl Tetrafluorobutanedioate, Tetrafluorosuccinic Acid Dimethyl Ester, Dimethyl Perfluorosuccinate

D3588-5G D3588-1G
TCD3588-5GEA 109.02 CAD
TCD3588-5G TCD3588-1G
Dimethyl tetrafluorosuccinate ≥95.0%
Dimethyl tetrafluorosuccinate

CAS Number: 356-36-5
MDL Number: MFCD00043947
Molecular Formula: C6H6F4O4
Molecular Weight: 218.10
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 174
Specific Gravity (20/20): 1.41


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Dimethyl Perfluorosuccinate, Perfluorosuccinic Acid Dimethyl Ester, Tetrafluorosuccinic Acid Dimethyl Ester, Dimethyl Tetrafluorobutanedioate, Tetrafluorobutanedioic Acid Dimethyl Ester

Formula: C₆H₆F₄O₄
Boiling Pt: 174 °C
Density: 1.41 (20 °C)
MDL Number: MFCD00043947
CAS Number: 356-36-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76f1dddf960bff49f1c82dd07f09dbb9243f364e&vid=76f1dddf960bff49f1c82dd07f09dbb9243f364e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR