5-Fluoro-5'-deoxyuridine (Doxifluridine) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D3579-1G D3579-5G
TCD3579-1GEA 256.28 CAD
TCD3579-1G TCD3579-5G
5-Fluoro-5'-deoxyuridine (Doxifluridine) ≥98.0% (by HPLC, titration analysis)
5-Fluoro-5'-deoxyuridine (Doxifluridine)

CAS Number: 3094-09-5
MDL Number: MFCD00866530
Molecular Formula: C9H11FN2O5
Molecular Weight: 246.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 189
Specific rotation [a]20/D: 20 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Antitumor Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: 5'-dFUrd, Doxifluridine, 1-(5-Deoxy-beta-D-ribofuranosyl)-5-fluorouracil

Formula: C₉H₁₁FN₂O₅
Melting Pt: 189 °C
MDL Number: MFCD00866530
CAS Number: 3094-09-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b63c95683f919c9083aa5a677a75d24879b5784c&vid=b63c95683f919c9083aa5a677a75d24879b5784c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR