Dimethyl-4-hydroxyisophthalate ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D3562-1G D3562-5G
TCD3562-1GEA 110.6 CAD
TCD3562-1G TCD3562-5G
Dimethyl-4-hydroxyisophthalate ≥97.0% (by GC, titration analysis)
Dimethyl-4-hydroxyisophthalate

CAS Number: 5985-24-0
MDL Number: MFCD00152444
Molecular Formula: C10H10O5
Molecular Weight: 210.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 97


Other Product Names / Synonyms: 4-Hydroxyisophthalic Acid Dimethyl Ester

Formula: C₁₀H₁₀O₅
Melting Pt: 97 °C
MDL Number: MFCD00152444
CAS Number: 5985-24-0

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR