Dimethyl hexadecanedioate ≥95.0%
Supplier: TCI America
D3554-25G
D3554-5G
TCD3554-25GEA
1342.21
CAD
TCD3554-25G
TCD3554-5G
Dimethyl hexadecanedioate ≥95.0%
Dimethyl hexadecanedioate
CAS Number: 19102-90-0
MDL Number: MFCD00039806
Molecular Formula: C18H34O4
Molecular Weight: 314.47
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 172
Melting point (°C): 53
Other Product Names / Synonyms: Hexadecanedioic Acid Dimethyl Ester
Formula:
C₁₈H₃₄O₄ Boiling Pt: 172 °C Melting Pt: 53 °C |
MDL Number:
MFCD00039806 CAS Number: 19102-90-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...