4,10-Dioxatricyclo[5.2.1.0(2,6)]decan-8-en-3-one ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,7-Epoxy-3a,4,7,7a-tetrahydroisobenzofuran-1(3H)-one

D3552-1G D3552-5G
TCD3552-1GEA 137.46 CAD
TCD3552-1G TCD3552-5G
4,10-Dioxatricyclo[5.2.1.0(2,6)]decan-8-en-3-one ≥97.0% (by HPLC)
4,10-Dioxatricyclo[5.2.1.0(2,6)]decan-8-en-3-one

CAS Number: 72150-22-2
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 93


Other Product Names / Synonyms: 4,7-Epoxy-3a,4,7,7a-tetrahydroisobenzofuran-1(3H)-one

Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 93 °C
CAS Number: 72150-22-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2de985a5bb7ecfc38205731019adb79030bcd61e&vid=2de985a5bb7ecfc38205731019adb79030bcd61e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR