Dimethyl (1-diazo-2-oxopropyl)phosphonate ≥97.0% (by HPLC)

Supplier: TCI America

D3546-1G D3546-5G
TCD3546-1GEA 497.42 CAD
TCD3546-1G TCD3546-5G
Dimethyl (1-diazo-2-oxopropyl)phosphonate ≥97.0% (by HPLC)
Dimethyl (1-diazo-2-oxopropyl)phosphonate

CAS Number: 90965-06-3
MDL Number: MFCD07368360
Molecular Formula: C5H9N2O4P
Molecular Weight: 192.11
Purity/Analysis Method: >97.0% (HPLC)
Form: Clear Liquid
Specific Gravity (20/20): 1.28
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Carbon Homologation / Degradation Reaction, Click Chemistry
Other Product Names / Synonyms: (1-Diazo-2-oxopropyl)phosphonic Acid Dimethyl Ester, Dimethyl 1-Diazoacetonylphosphonate, 1-Diazoacetonylphosphonic Acid Dimethyl Ester, Ohira-Bestmann Reagent

Formula: C₅H₉N₂O₄P
Density: 1.28 (20 °C)
MDL Number: MFCD07368360
CAS Number: 90965-06-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecd8f383d374107e604a22c2353695dbdc5920ca&vid=ecd8f383d374107e604a22c2353695dbdc5920ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR