[N-[ɑ-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel ≥95.0%

Supplier: TCI America

D3543-1G D3543-100MG
TCD3543-1GEA 337.17 CAD
TCD3543-1G TCD3543-100MG
[N-[ɑ-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel ≥95.0%
[N-[alpha-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel

CAS Number: 847654-16-4
Molecular Formula: C25H31N3NiO3
Molecular Weight: 480.23
Purity/Analysis Method: >95.0% (T)
Form: Crystal

Formula: C₂₅H₃₁N₃NiO₃
MW: 480.23 g/mol
CAS Number: 847654-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d95e3e554255a32019bcc88d38bfcc21f0295d80&vid=d95e3e554255a32019bcc88d38bfcc21f0295d80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR