Dimethyl octadecanedioate ≥96.0%

Supplier: TCI America

D3540-25G D3540-5G
TCD3540-25GEA 923.98 CAD
TCD3540-25G TCD3540-5G
Dimethyl octadecanedioate ≥96.0%
Dimethyl octadecanedioate

CAS Number: 1472-93-1
MDL Number: MFCD00673461
Molecular Formula: C20H38O4
Molecular Weight: 342.52
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 207
Melting point (°C): 60


Other Product Names / Synonyms: Octadecanedioic Acid Dimethyl Ester

Formula: C₂₀H₃₈O₄
Boiling Pt: 207 °C
Melting Pt: 60 °C
MDL Number: MFCD00673461
CAS Number: 1472-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5932566dcab62812ffe7625526bc88ffdf8819db&vid=5932566dcab62812ffe7625526bc88ffdf8819db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR