4-(N,N-Diphenylamino)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
4-(Diphenylamino)benzeneboronic acid, 4-(Diphenylamino)phenylboronic acid
D3537-1G
D3537-5G
TCD3537-1GEA
45.82
CAD
TCD3537-1G
TCD3537-5G
4-(N,N-Diphenylamino)phenylboronic acid (contains varying amounts of Anhydride)
4-(N,N-Diphenylamino)phenylboronic acid
CAS Number: 201802-67-7
MDL Number: MFCD06798117
Molecular Formula: C18H16BNO2
Molecular Weight: 289.14
Form: Crystal
Color: White
Melting point (°C): 228
Other Product Names / Synonyms: Triphenylamine-4-boronic Acid
Formula:
C₁₈H₁₆BNO₂ MW: 289.14 g/mol Melting Pt: 228 °C |
MDL Number:
MFCD06798117 CAS Number: 201802-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...