4-(Dimethylamino)butyraldehyde dimethyl acetal ≥98.0%
Supplier: TCI America
Synonyms:
4,4-Dimethoxy-N,N-dimethylbutylamine
D3535-5G
D3535-25G
TCD3535-5GEA
150.1
CAD
TCD3535-5G
TCD3535-25G
4-(Dimethylamino)butyraldehyde dimethyl acetal ≥98.0%
4-(Dimethylamino)butyraldehyde dimethyl acetal
CAS Number: 19718-92-4
MDL Number: MFCD06411126
Molecular Formula: C8H19NO2
Molecular Weight: 161.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 54
Specific Gravity (20/20): 0.89
Other Product Names / Synonyms: 4,4-Dimethoxy-N,N-dimethylbutylamine
Formula:
C₈H₁₉NO₂ MW: 161.25 g/mol Boiling Pt: 54 °C Density: 0.89 (20 °C) |
MDL Number:
MFCD06411126 CAS Number: 19718-92-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...