3,5-Dimethoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

D3513-1G D3513-5G D3513-25G
TCD3513-1GEA 53.72 CAD
TCD3513-1G TCD3513-5G TCD3513-25G
3,5-Dimethoxyphenylboronic acid (contains varying amounts of Anhydride)
3,5-Dimethoxyphenylboronic acid

CAS Number: 192182-54-0
MDL Number: MFCD03095127
Molecular Formula: C8H11BO4
Molecular Weight: 181.98
Form: Crystal
Color: White
Melting point (°C): 202


Other Product Names / Synonyms: 3,5-Dimethoxybenzeneboronic Acid

Formula: C₈H₁₁BO₄
Melting Pt: 202 °C
MDL Number: MFCD03095127
CAS Number: 192182-54-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4968db0ca5e9e6467bc1e900cf58be7698fc81c8&vid=4968db0ca5e9e6467bc1e900cf58be7698fc81c8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR