Diethyl pyrazole-3,5-dicarboxylate ≥98.0%
Supplier: TCI America
D3507-1G
D3507-5G
TCD3507-1GEA
63.2
CAD
TCD3507-1G
TCD3507-5G
Diethyl pyrazole-3,5-dicarboxylate ≥98.0%
Diethyl pyrazole-3,5-dicarboxylate
CAS Number: 37687-24-4
MDL Number: MFCD00152167
Molecular Formula: C9H12N2O4
Molecular Weight: 212.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 55
Other Product Names / Synonyms: Pyrazole-3,5-dicarboxylic Acid Diethyl Ester
Formula:
C₉H₁₂N₂O₄ Melting Pt: 55 °C |
MDL Number:
MFCD00152167 CAS Number: 37687-24-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...