2,5-Dimethyl-4-methoxy-3(2H)-furanone ≥97.0%
Supplier: TCI America
D3499-25G
D3499-5G
TCD3499-25GEA
99.54
CAD
TCD3499-25G
TCD3499-5G
2,5-Dimethyl-4-methoxy-3(2H)-furanone ≥97.0%
2,5-Dimethyl-4-methoxy-3(2H)-furanone
CAS Number: 4077-47-8
MDL Number: MFCD00209504
Molecular Formula: C7H10O3
Molecular Weight: 142.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 63
Flash Point (°C): 75
Specific Gravity (20/20): 1.10
Formula:
C₇H₁₀O₃ Boiling Pt: 63 °C Density: 1.10 (20 °C) Flash Pt: 75 °C |
MDL Number:
MFCD00209504 CAS Number: 4077-47-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...