2,5-Diacetoxyhexane ≥98.0%

Supplier: TCI America

D3496-25G D3496-5G
TCD3496-25GEA 567.22 CAD
TCD3496-25G TCD3496-5G
2,5-Diacetoxyhexane ≥98.0%
2,5-Diacetoxyhexane

CAS Number: 10299-35-1
Molecular Formula: C10H18O4
Molecular Weight: 202.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 2,5-Hexanediol Diacetate

Formula: C₁₀H₁₈O₄
Boiling Pt: 122 °C
Density: 1.00 (20 °C)
CAS Number: 10299-35-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5aa17af7225daf78e6a1f05a89de73b1eafcc3b3&vid=5aa17af7225daf78e6a1f05a89de73b1eafcc3b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR