5,6-Dichloro-2-methylbenzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3488-5G D3488-1G
TCD3488-5GEA 521.4 CAD
TCD3488-5G TCD3488-1G
5,6-Dichloro-2-methylbenzimidazole ≥98.0% (by GC, titration analysis)
5,6-Dichloro-2-methylbenzimidazole

CAS Number: 6478-79-1
MDL Number: MFCD00276973
Molecular Formula: C8H6Cl2N2
Molecular Weight: 201.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 248

Formula: C₈H₆Cl₂N₂
MW: 201.0500 g/mol
Melting Pt: 248 °C
MDL Number: MFCD00276973
CAS Number: 6478-79-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5620837ffc41e0cc07a0386ef9bd0d63019f42c2&vid=5620837ffc41e0cc07a0386ef9bd0d63019f42c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR