4-(Dimethylamino)benzyl alcohol ≥98.0%
Supplier: TCI America
Synonyms:
4-Hydroxymethyl-N,N-dimethylaniline
D3482-25G
D3482-5G
TCD3482-25GEA
1507.32
CAD
TCD3482-25G
TCD3482-5G
4-(Dimethylamino)benzyl alcohol ≥98.0%
4-(Dimethylamino)benzyl alcohol
CAS Number: 1703-46-4
MDL Number: MFCD03840449
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 123
Specific Gravity (20/20): 1.07
Storage Temperature: <0°C
Other Product Names / Synonyms: 4-Hydroxymethyl-N,N-dimethylaniline
Formula:
C₉H₁₃NO MW: 151.21 g/mol Boiling Pt: 123 °C Density: 1.07 (20 °C) |
MDL Number:
MFCD03840449 CAS Number: 1703-46-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...