1,3-Di-tert-butylimidazol-2-ylidene ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
ItBu, 1,3-Di-t-butylimidazol-2-ylidene
D3472-1G
D3472-5G
TCD3472-1GEA
359.01
CAD
TCD3472-1G
TCD3472-5G
1,3-Di-tert-butylimidazol-2-ylidene ≥98.0% (by titrimetric analysis)
1,3-Di-tert-butylimidazol-2-ylidene
CAS Number: 157197-53-0
MDL Number: MFCD03788921
Molecular Formula: C11H20N2
Molecular Weight: 180.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands, N-Heterocyclic Carbene Ligands ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: ItBu
Formula:
C₁₁H₂₀N₂ MW: 180.30 g/mol |
MDL Number:
MFCD03788921 CAS Number: 157197-53-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...