Dimethyl-4-acetoxyisophthalate ≥98.0%

Supplier: TCI America

A1226-1G A1226-5G
TCA1226-1GEA 164.32 CAD
TCA1226-1G TCA1226-005G
Dimethyl-4-acetoxyisophthalate ≥98.0%
Dimethyl-4-acetoxyisophthalate

CAS Number: 71932-29-1
MDL Number: MFCD00143241
Molecular Formula: C12H12O6
Molecular Weight: 252.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 96


Other Product Names / Synonyms: 4-Acetoxyisophthalic Acid Dimethyl Ester

Formula: C₁₂H₁₂O₆
Melting Pt: 96 °C
MDL Number: MFCD00143241
CAS Number: 71932-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98388d7556e1db8907f46ad71475ec31a3391371&vid=98388d7556e1db8907f46ad71475ec31a3391371&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR