Dipropylene glycol diacrylate ≥80.0% stabilised

Supplier: TCI America

D3470-500G D3470-25G
TCD3470-500GEA 252.96 CAD
TCD3470-500G TCD3470-25G TCD3470-100G
Dipropylene glycol diacrylate ≥80.0% stabilised
Dipropylene glycol diacrylate

(stabilized with MEHQ)
CAS Number: 57472-68-1
Molecular Formula: C12H18O5
Molecular Weight: 242.27
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.05

Formula: C₁₂H₁₈O₅
MW: 242.27 g/mol
Density: 1.05 (20 °C)
MDL Number: MFCD00242743
CAS Number: 57472-68-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9c5caaedefb21d89dee884998d87fd1cab404a7&vid=b9c5caaedefb21d89dee884998d87fd1cab404a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR